Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

476382

Sigma-Aldrich

N,N-Dimethylvinylbenzylamine, mixture of isomers

97%

Synonym(s):

(Dimethylaminomethyl)styrene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2C=CHC6H4CH2N(CH3)2
CAS Number:
Molecular Weight:
161.24
EC Number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

assay

97%

refractive index

n20/D 1.539 (lit.)

density

0.922 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

CN(C)Cc1ccc(C=C)cc1.CN(C)Cc2cccc(C=C)c2.CN(C)Cc3ccccc3C=C

InChI

1S/C11H15N/c1-4-10-5-7-11(8-6-10)9-12(2)3/h4-8H,1,9H2,2-3H3

InChI key

XQBHAZDVLGNSOJ-UHFFFAOYSA-N

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

199.4 °F - closed cup

flash_point_c

93 °C - closed cup

ppe

Eyeshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service