Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

47603

Sigma-Aldrich

Fmoc-Cys(Acm)-OH

≥95.0% (sum of enantiomers, HPLC)

Synonym(s):

Nα-Fmoc-S-acetaminomethyl-L-cysteine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H22N2O5S
CAS Number:
Molecular Weight:
414.47
Beilstein/REAXYS Number:
4725388
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.26

assay

≥95.0% (sum of enantiomers, HPLC)

optical activity

[α]/D -33.0±2.0°, c = 1 in methanol

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

application(s)

peptide synthesis

functional group

Fmoc

storage temp.

2-8°C

SMILES string

CC(=O)NCSC[C@H](NC(=O)OCC1c2ccccc2-c3ccccc13)C(O)=O

InChI

1S/C21H22N2O5S/c1-13(24)22-12-29-11-19(20(25)26)23-21(27)28-10-18-16-8-4-2-6-14(16)15-7-3-5-9-17(15)18/h2-9,18-19H,10-12H2,1H3,(H,22,24)(H,23,27)(H,25,26)/t19-/m0/s1

InChI key

CSMYOORPUGPKAP-IBGZPJMESA-N

Looking for similar products? Visit Product Comparison Guide

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Enrica Calce et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 24(23), 6231-6238 (2018-02-20)
An efficient and rapid procedure for synthesizing S-linked glycopeptides is reported. The approach uses activated molecular sieves as a base to promote the selective S-alkylation of readily prepared cysteine-containing peptides, upon reaction of appropriate glycosyl halides. Considering the very mild
Kusum Vats et al.
Journal of peptide science : an official publication of the European Peptide Society, 25(7), e3192-e3192 (2019-07-17)
The present study describes modification of asparagine-glycine-arginine (NGR) peptide at N-terminally and C-terminally by introduction of a tridentate chelating scaffold via click chemistry reaction. The N-terminal and C-terminal modified peptides were radiometalated with [99m Tc(CO)3 ]+ precursor. The influence of
S N McCurdy
Peptide research, 2(1), 147-152 (1989-01-01)
Fmoc-Cys(t-Bu)-OH, Fmoc-Cys(Acm)-OH, and Fmoc-Cys(Trt)-OH exhibit excellent synthesis characteristics when used in Fmoc solid phase peptide synthesis on the Applied Biosystems Model 431A peptide synthesizer. The actual 5% scavenger mixture will vary according to the particular amino acid residues present. As
Emma J Ste Marie et al.
Journal of peptide science : an official publication of the European Peptide Society, 24(11), e3130-e3130 (2018-10-26)
Triisopropylsilane (TIS), a hindered hydrosilane, has long been utilized as a cation scavenger for the removal of amino acid protecting groups during peptide synthesis. However, its ability to actively remove S-protecting groups by serving as a reductant has largely been
Fan Gao et al.
Small (Weinheim an der Bergstrasse, Germany), 14(37), e1802403-e1802403 (2018-08-22)
Chemotherapy is well recognized to induce immune responses during some chemotherapeutic drugs-mediated tumor eradication. Here, a strategy involving blocking programmed cell death protein 1 (PD-1) to enhance the chemotherapeutic effect of a doxorubicin nanoprodrug HA-Psi-DOX is proposed and the synergetic

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service