Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

47565

Sigma-Aldrich

Fmoc-Tyr(Bzl)-OH

≥98.0% (HPLC)

Synonym(s):

Fmoc-O-benzyl-L-tyrosine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C31H27NO5
CAS Number:
Molecular Weight:
493.55
Beilstein/REAXYS Number:
4341195
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

assay

≥98.0% (HPLC)

optical activity

[α]20/D −16.0±2.5°, c = 1% in DMF

mp

157-161 °C

application(s)

peptide synthesis

functional group

Fmoc

storage temp.

2-8°C

SMILES string

OC(=O)[C@H](Cc1ccc(OCc2ccccc2)cc1)NC(=O)OCC3c4ccccc4-c5ccccc35

InChI

1S/C31H27NO5/c33-30(34)29(18-21-14-16-23(17-15-21)36-19-22-8-2-1-3-9-22)32-31(35)37-20-28-26-12-6-4-10-24(26)25-11-5-7-13-27(25)28/h1-17,28-29H,18-20H2,(H,32,35)(H,33,34)/t29-/m0/s1

InChI key

REHSJSKPWIOKIJ-LJAQVGFWSA-N

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chloé Guilbaud-Chéreau et al.
ACS applied materials & interfaces, 11(14), 13147-13157 (2019-03-14)
Molecular gels formed by the self-assembly of low-molecular-weight gelators have received increasing interest because of their potential applications in drug delivery. In particular, the ability of peptides and amino acids to spontaneously self-assemble into three-dimensional fibrous network has been exploited

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service