Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

475386

Sigma-Aldrich

N,N′,N′′-Tri-tert-butylsilanetriamine

99.99% trace metals basis

Synonym(s):

Tris(tert -butylamino)silane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HSi(HNC(CH3)3)3
CAS Number:
Molecular Weight:
245.48
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:

assay

99.99% trace metals basis

form

liquid

refractive index

n20/D 1.4289 (lit.)

bp

76-80 °C/0.3 mmHg (lit.)

density

0.827 g/mL at 25 °C (lit.)

SMILES string

CC(C)(C)N[SiH](NC(C)(C)C)NC(C)(C)C

InChI

1S/C12H31N3Si/c1-10(2,3)13-16(14-11(4,5)6)15-12(7,8)9/h13-16H,1-9H3

InChI key

UGJHADISJBNSFP-UHFFFAOYSA-N

pictograms

FlameCorrosion

signalword

Danger

hcodes

Hazard Classifications

Flam. Liq. 3 - Skin Corr. 1B

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

129.0 °F - closed cup

flash_point_c

53.9 °C - closed cup

ppe

Faceshields, Gloves, Goggles, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Synthesis of Melting Gels Using Mono-Substituted and Di-Substituted Alkoxysiloxanes

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service