Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

474843

Sigma-Aldrich

2,5-Difluorophenyl isothiocyanate

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
F2C6H3NCS
CAS Number:
Molecular Weight:
171.17
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

assay

97%

refractive index

n20/D 1.602 (lit.)

bp

205 °C (lit.)

density

1.338 g/mL at 25 °C (lit.)

SMILES string

Fc1ccc(F)c(c1)N=C=S

InChI

1S/C7H3F2NS/c8-5-1-2-6(9)7(3-5)10-4-11/h1-3H

InChI key

QVIDXDRFMDPVLA-UHFFFAOYSA-N

pictograms

Corrosion

signalword

Danger

hcodes

Hazard Classifications

Skin Corr. 1B

Storage Class

8A - Combustible corrosive hazardous materials

wgk_germany

WGK 3

flash_point_f

197.6 °F - closed cup

flash_point_c

92 °C - closed cup

ppe

Faceshields, Gloves, Goggles, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service