Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

474401

Sigma-Aldrich

2,2,3,3,4,4,5,5-Octafluoropentyl acrylate

97%, contains 100 ppm monomethyl ether hydroquinone as inhibitor

Synonym(s):

OFPA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2C=CHCO2CH2(CF2)3CHF2
CAS Number:
Molecular Weight:
286.12
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

assay

97%

contains

100 ppm monomethyl ether hydroquinone as inhibitor

refractive index

n20/D 1.349 (lit.)

bp

122 °C/140 mmHg (lit.)

density

1.488 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

FC(F)C(F)(F)C(F)(F)C(F)(F)COC(=O)C=C

InChI

1S/C8H6F8O2/c1-2-4(17)18-3-6(11,12)8(15,16)7(13,14)5(9)10/h2,5H,1,3H2

InChI key

WISUNKZXQSKYMR-UHFFFAOYSA-N

pictograms

Exclamation markEnvironment

signalword

Warning

Hazard Classifications

Aquatic Chronic 2 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

10 - Combustible liquids

wgk_germany

WGK 2

flash_point_f

161.6 °F - closed cup

flash_point_c

72 °C - closed cup

ppe

Eyeshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yisheng He et al.
Hepatology (Baltimore, Md.), 74(1), 264-280 (2021-01-20)
Mutational signature analyses are an effective tool in identifying cancer etiology. Humans are frequently exposed to pyrrolizidine alkaloids (PAs), the most common carcinogenic phytotoxins widely distributed in herbal remedies and foods. However, due to the lack of human epidemiological data
Christian Hoffmann et al.
Biotechnology progress, 33(5), 1267-1277 (2017-07-20)
Efficient immobilization of enzymes on support surfaces requires an exact match between the surface chemistry and the specific enzyme. A successful match would normally be identified through time consuming screening of conventional resins in multiple experiments testing individual immobilization strategies.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service