Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

47410

Sigma-Aldrich

4-Fluorophthalonitrile

≥99.0%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H3FN2
CAS Number:
Molecular Weight:
146.12
Beilstein/REAXYS Number:
2443236
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

assay

≥99.0%

form

solid

mp

100-104 °C

SMILES string

Fc1ccc(C#N)c(c1)C#N

InChI

1S/C8H3FN2/c9-8-2-1-6(4-10)7(3-8)5-11/h1-3H

InChI key

QQEKYCCJLSRLEC-UHFFFAOYSA-N

General description

4-Fluorophthalonitrile is an aryl fluorinated building block.

Application

4-Fluorophthalonitrile may be used in the synthesis of 2,29,20,2-tetrafluoropthalocyanito zinc(II).[1]

pictograms

Skull and crossbones

signalword

Danger

hcodes

Hazard Classifications

Acute Tox. 2 Oral

Storage Class

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Faceshields, Gloves, type P3 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Darya Klyamer et al.
Sensors (Basel, Switzerland), 20(7) (2020-04-03)
In this work, thin films of vanadyl phthalocyanines (VOPc and VOPcF4) are studied as active layers for the detection of gaseous ammonia and hydrogen. The effect of F-substituents on the structural features of vanadyl phthalocyanine films and their sensor response
Woll C.
Physical and Chemical Aspects of Organic Electronics, 39-39 (2009)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service