Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

471143

Sigma-Aldrich

Tetrakis[1-[[4-alkyl(C11-C13)phenyl]sulfonyl]-(2R)-pyrrolidinecarboxylate]dirhodium(II)

Synonym(s):

Rh2(R-DOSP)4

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352005

form

solid

optical activity

[α]22/D +165°, c = 0.1 in chloroform

reaction suitability

reaction type: Cyclopropanations

mp

224-230 °C (lit.)

storage temp.

2-8°C

Application

Catalyst for asymmetric cyclopropanations.[1][2][3][4]

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Davies, H.M.L. Hutcheson, D.K.
Tetrahedron Letters, 34, 7243-7243 (1993)
Davies, H.M.L. et al.
Tetrahedron Letters, 38, 1741-1741 (1997)
Davies, H.M.L. et al.
Journal of the American Chemical Society, 118, 6897-6897 (1996)
Davies, H.M.L. et al.
Tetrahedron Letters, 37, 4133-4133 (1996)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service