All Photos(1)
About This Item
Linear Formula:
ClC6H4OSi(CH3)3
CAS Number:
Molecular Weight:
200.74
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
assay
97%
refractive index
n20/D 1.494 (lit.)
bp
202 °C (lit.)
density
1.038 g/mL at 25 °C (lit.)
SMILES string
C[Si](C)(C)Oc1ccccc1Cl
InChI
1S/C9H13ClOSi/c1-12(2,3)11-9-7-5-4-6-8(9)10/h4-7H,1-3H3
InChI key
NZLHRQHCIXYIMM-UHFFFAOYSA-N
signalword
Warning
hcodes
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Storage Class
10 - Combustible liquids
wgk_germany
WGK 3
flash_point_f
170.6 °F - closed cup
flash_point_c
77 °C - closed cup
ppe
Eyeshields, Gloves, type ABEK (EN14387) respirator filter
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service