Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

470228

Sigma-Aldrich

1-Dimethylamino-2-propanol solution

77 wt. % in H2O

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2NCH2CH(OH)CH3
CAS Number:
Molecular Weight:
103.16
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

concentration

77 wt. % in H2O

refractive index

n20/D 1.42

bp

96 °C

density

0.913 g/mL at 25 °C

SMILES string

CC(O)CN(C)C

InChI

1S/C5H13NO/c1-5(7)4-6(2)3/h5,7H,4H2,1-3H3

Inchi Key

NCXUNZWLEYGQAH-UHFFFAOYSA-N

replaced by

Product No.
Description
Pricing

signalword

Danger

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1B

Storage Class

3 - Flammable liquids

wgk_germany

WGK 1

flash_point_f

95.9 - 99.0 °F - closed cup

flash_point_c

35.5 - 37.2 °C - closed cup

ppe

Faceshields, Gloves, Goggles, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kinetics of carbon dioxide (CO 2) with ethylenediamine, 3-amino-1-propanol in methanol and ethanol, and with 1-dimethylamino-2-propanol and 3-dimethylamino-1-propanol in water using stopped-flow technique
Kadiwala S, et al.
Chemical Engineering Journal, 179, 262-271 (2012)
Soluble Ni II alkoxides based on dimethylaminoisopropoxide ligands: molecular structure of [Li (Pr i OH) Ni (η 2-OR)2 Cl]2 and of cis-NiCl2 (ROH)2 (R= CHMeCH2NMe2).
Hubert-Pfalzgraf LG, et al.
Polyhedron, 16(24), 4197-4203 (1997)
S A Naujokaitis et al.
Journal of pharmaceutical sciences, 73(1), 34-39 (1984-01-01)
The structure-activity relationships of 2-dimethylaminoethanol and its analogues as protectors against mechlorethamine cytotoxicity and as inhibitors of choline uptake were evaluated. Of a series of inhibitors and protectors, 2-dimethylaminoethanol was the most potent inhibitor of choline uptake and the most
Synthesis and molecular structures of cobalt (II) ?-diketonate complexes as new MOCVD precursors for cobalt oxide films.
Pasko S, et al.
Polyhedron, 23(5), 735-741 (2004)
Zhaopin Bai et al.
Inorganic chemistry, 49(19), 9005-9011 (2010-09-04)
A new pathway by means of transetherification has been developed to synthesize novel unsymmetrical 2,3,9,10,16,17,23-heptakis(alkoxyl)-24-mono(dimethylaminoalkoxyl)phthalocyanine compounds. Cyclic tetramerization of 4,5-di(alkoxyl)phthalonitrile in refluxing dimethylamino-alcohol with high boiling point such as dimethylaminoethanol (DMAE) and 1-dimethylamino-2-propanol in the presence of lithium and pyrazino[2,3-f][1,10]phenanthroline-2,3-dicarbonitrile

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service