Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

470074

Sigma-Aldrich

4-(Trifluoromethylthio)aniline

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CF3SC6H4NH2
CAS Number:
Molecular Weight:
193.19
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

assay

97%

refractive index

n20/D 1.528 (lit.)

bp

102-103 °C/8 mmHg (lit.)

density

1.351 g/mL at 25 °C (lit.)

SMILES string

Nc1ccc(SC(F)(F)F)cc1

InChI

1S/C7H6F3NS/c8-7(9,10)12-6-3-1-5(11)2-4-6/h1-4H,11H2

InChI key

OHHHTUXVBNGOGI-UHFFFAOYSA-N

General description

4-(Trifluoromethylthio)aniline is a 4-substituted aniline derivative.[1]

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

224.6 °F - closed cup

flash_point_c

107 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Copper-Free Direct C-H Trifluoromethylation of Acetanilides with Sodium Trifluoromethanesulfinate.
Wu M, et al.
The Journal of Organic Chemistry, 79(19), 8984-8989 (2014)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service