Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

470007

Sigma-Aldrich

D-Histidinol dihydrochloride

98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H11N3O · 2HCl
CAS Number:
Molecular Weight:
214.09
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.22

assay

98%

optical activity

[α]20/D +3°, c = 1 in H2O

reaction suitability

reaction type: solution phase peptide synthesis

mp

193-195 °C (lit.)

application(s)

peptide synthesis

SMILES string

Cl.Cl.N[C@@H](CO)Cc1c[nH]cn1

InChI

1S/C6H11N3O.2ClH/c7-5(3-10)1-6-2-8-4-9-6;;/h2,4-5,10H,1,3,7H2,(H,8,9);2*1H/t5-;;/m1../s1

InChI key

FRCAFNBBXRWXQA-ZJIMSODOSA-N

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zhihong Zeng et al.
The Journal of biological chemistry, 286(1), 403-409 (2010-10-30)
Topoisomerase II (Top2) activity involves an intermediate in which the topoisomerase is covalently bound to a DNA double-strand break via a 5'-phosphotyrosyl bond. Although these intermediates are normally transient, they can be stabilized by antitumor agents that act as Top2

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service