Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

469467

Sigma-Aldrich

(S)-(−)-3-(Benzyloxycarbonyl)-4-oxazolidinecarboxylic acid

98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H13NO5
CAS Number:
Molecular Weight:
251.24
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

assay

98%

form

solid

optical activity

[α]20/D −92°, c = 1 in chloroform

optical purity

ee: 97% (HPLC)

mp

74-78 °C (lit.)

SMILES string

OC(=O)[C@@H]1COCN1C(=O)OCc2ccccc2

InChI

1S/C12H13NO5/c14-11(15)10-7-17-8-13(10)12(16)18-6-9-4-2-1-3-5-9/h1-5,10H,6-8H2,(H,14,15)/t10-/m0/s1

Inchi Key

XRRRGBIMHQARMF-JTQLQIEISA-N

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Proline analogues are promising candidates for tuning the biological, pharmaceutical, or physicochemical properties of naturally occuring, as well as de novo designed, linear, and, cyclic peptides.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service