Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

469033

Sigma-Aldrich

Pentylmagnesium chloride solution

2.0 M in THF

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)4MgCl
CAS Number:
Molecular Weight:
130.90
MDL number:
UNSPSC Code:
12352001
PubChem Substance ID:
NACRES:
NA.22

reaction suitability

reaction type: Grignard Reaction

concentration

2.0 M in THF

bp

65 °C

density

0.967 g/mL at 25 °C

Storage temp.

2-8°C

SMILES string

CCCCC[Mg]Cl

InChI

1S/C5H11.ClH.Mg/c1-3-5-4-2;;/h1,3-5H2,2H3;1H;/q;;+1/p-1

Inchi Key

OHLJPYMGJFINNA-UHFFFAOYSA-M

signalword

Danger

Hazard Classifications

Carc. 2 - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1B - STOT SE 3 - Water-react 2

target_organs

Respiratory system

supp_hazards

Storage Class

4.3 - Hazardous materials which set free flammable gases upon contact with water

wgk_germany

WGK 3

flash_point_f

1.0 °F - closed cup

flash_point_c

-17.2 °C - closed cup

ppe

Faceshields, Gloves, Goggles


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Oleg V Larionov et al.
Organic letters, 16(3), 864-867 (2014-01-15)
A one-step transformation of heterocyclic N-oxides to 2-alkyl-, aryl-, and alkenyl-substituted N-heterocycles is described. The success of this broad-scope methodology hinges on the combination of copper catalysis and activation by lithium fluoride or magnesium chloride. The utility of this method

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service