Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

467545

Sigma-Aldrich

4-tert-Butylcyclohexyl acrylate

90%

Synonym(s):

4-tert -Butylcyclohexyl acrylate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2C=CHCO2C6H10C(CH3)3
CAS Number:
Molecular Weight:
210.31
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:

Quality Level

assay

90%

refractive index

n20/D 1.464 (lit.)

density

1.108 g/mL at 25 °C (lit.)

SMILES string

CC(C)(C)C1CCC(CC1)OC(=O)C=C

InChI

1S/C13H22O2/c1-5-12(14)15-11-8-6-10(7-9-11)13(2,3)4/h5,10-11H,1,6-9H2,2-4H3

InChI key

LAIJAUHBAWLPCO-UHFFFAOYSA-N

pictograms

Exclamation markEnvironment

signalword

Warning

Hazard Classifications

Aquatic Chronic 2 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

215.6 °F - closed cup

flash_point_c

102 °C - closed cup

ppe

Eyeshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Antonio Conde-González et al.
Materials science & engineering. C, Materials for biological applications, 108, 110489-110489 (2020-01-12)
Three dimensional synthetic polymer scaffolds have remarkable chemical and mechanical tunability in addition to biocompatibility. However, the chemical and physical space is vast in view of the number of variables that can be altered e.g. chemical composition, porosity, pore size

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service