About This Item
Recommended Products
assay
95%
form
solid
mp
221 °C (dec.) (lit.)
storage temp.
−20°C
SMILES string
[O-][N+](=O)c1cccc2n[se]nc12
InChI
1S/C6H3N3O2Se/c10-9(11)5-3-1-2-4-6(5)8-12-7-4/h1-3H
InChI key
PUKFAFPOBCMYAI-UHFFFAOYSA-N
General description
Application
signalword
Danger
hcodes
Hazard Classifications
Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - STOT RE 2
Storage Class
6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
dust mask type N95 (US), Eyeshields, Faceshields, Gloves, type P2 (EN 143) respirator cartridges
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service