Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

467073

Sigma-Aldrich

(3aS,3′aS,8aR,8′aR)-2,2′-Methylenebis[3a,8a-dihydro-8H-indeno[1,2-d]oxazole]

98%

Synonym(s):

[3aS-[2(3′aR*,8′aS*),3′aα,8′aα]]-(−)-2,2′-Methylenebis[3a,8a-dihydro-8H-indeno[1,2-d]oxazole]

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H18N2O2
CAS Number:
Molecular Weight:
330.38
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

assay

98%

form

solid

optical activity

[α]22/D −355°, c = 3.7 in chloroform

mp

215-219 °C (lit.)

functional group

ether

SMILES string

C1[C@H]2OC(CC3=N[C@@H]4[C@H](Cc5ccccc45)O3)=N[C@@H]2c6ccccc16

InChI

1S/C21H18N2O2/c1-3-7-14-12(5-1)9-16-20(14)22-18(24-16)11-19-23-21-15-8-4-2-6-13(15)10-17(21)25-19/h1-8,16-17,20-21H,9-11H2/t16-,17-,20+,21+/m1/s1

InChI key

BDHSVQLSNIGJNC-JYWFKMLOSA-N

Application

(3aS,3′aS,8aR,8′aR)-2,2′-Methylenebis[3a,8a-dihydro-8H-indeno[1,2-d]oxazole] can be used as a chiral ligand:
  • In the preparation of oxazole α-hydroxy esters through intermolecular Alder-Ene reaction.[1]
  • In the atom transfer radical polymerization reactions of methyl methacrylate.[2]
  • In the copper-catalyzed synthesis of isoxazolidine derivatives by reacting nitrone with α,β-unsaturated acyl phosphonate.[3]

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sequential Au/Cu Catalysis: A Two Catalyst One-Pot Protocol for the Enantioselective Synthesis of Oxazole ?-Hydroxy Esters via Intramolecular Cyclization/Intermolecular Alder-Ene Reaction
Nalivela KS, et al.
Advanced Synthesis & Catalysis, 360(11), 2183-2190 (2018)
Highly Efficient Atom Transfer Radical Polymerization System Based on the SaBOX/Copper Catalyst
Chen Z, et al.
Macromolecules, 52(24), 9792?9798-9792?9798 (2019)
Cu (II)-catalyzed enantioselective 1, 3-dipolar cycloaddition of nitrones with α,β-unsaturated acyl phosphonates
Xie L, et al.
Tetrahedron, 73(20), 2923-2930 (2017)
Sequential Au/Cu Catalysis: A Two Catalyst One-Pot Protocol for the Enantioselective Synthesis of Oxazole ?-Hydroxy Esters via Intramolecular Cyclization/Intermolecular Alder-Ene Reaction
Nalivela KS, et al.
advanced synthesis and catalysis, 360(11), 2183-2190 (2018)

Articles

BOX ligand complex enhances catalytic enantioselective aziridination of styrene, with phenyl substituents proving superior over t-Bu groups.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service