Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

466107

Sigma-Aldrich

Magnesium iodide

powder, 99.998% trace metals basis

Synonym(s):

Magnesium diiodide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
MgI2
CAS Number:
Molecular Weight:
278.11
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
assay:
99.998% trace metals basis
form:
powder

Quality Level

assay

99.998% trace metals basis

form

powder

mp

637 °C (dec.) (lit.)

density

4.43 g/mL at 25 °C (lit.)

SMILES string

I[Mg]I

InChI

1S/2HI.Mg/h2*1H;/q;;+2/p-2

InChI key

BLQJIBCZHWBKSL-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

application

When reacted with cyclic sulfates of vic-diols, produces the corresponding olefins in excellent yields.[1]

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jang, D.O. Joo, Y.H.
Synthetic Communications, 28, 871-871 (1998)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service