Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

465674

Sigma-Aldrich

N-(tert-Butoxycarbonyl)glycine methyl ester

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3COCONHCH2CO2CH3
CAS Number:
Molecular Weight:
189.21
MDL number:
UNSPSC Code:
12352200
eCl@ss:
32160406
PubChem Substance ID:
NACRES:
NA.22

assay

97%

reaction suitability

reaction type: solution phase peptide synthesis

refractive index

n20/D 1.437 (lit.)

bp

190 °C (lit.)

density

1.079 g/mL at 25 °C

application(s)

peptide synthesis

SMILES string

COC(=O)CNC(=O)OC(C)(C)C

InChI

1S/C8H15NO4/c1-8(2,3)13-7(11)9-5-6(10)12-4/h5H2,1-4H3,(H,9,11)

InChI key

PHUZOEOLWIHIKH-UHFFFAOYSA-N

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

228.2 °F - closed cup

flash_point_c

109 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service