Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

464015

Sigma-Aldrich

(S)-(+)-2-Benzyl-1-(p-tolylsulfonyl)aziridine

98%

Synonym(s):

(S)-2-Benzyl-1-tosylaziridine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H17NO2S
CAS Number:
Molecular Weight:
287.38
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

assay

98%

optical activity

[α]20/D +8.8°, c = 1.3 in toluene

mp

92-94 °C (lit.)

functional group

phenyl
sulfonamide

SMILES string

Cc1ccc(cc1)S(=O)(=O)N2C[C@@H]2Cc3ccccc3

InChI

1S/C16H17NO2S/c1-13-7-9-16(10-8-13)20(18,19)17-12-15(17)11-14-5-3-2-4-6-14/h2-10,15H,11-12H2,1H3/t15-,17?/m0/s1

InChI key

ISURUORMAKCTFF-MYJWUSKBSA-N

Application

(S)-(+)-2-Benzyl-1-(p-tolylsulfonyl)aziridine can be used:
  • To prepare tert-butyl(S)-6-((4-methylphenyl)sulfonamido)-3-oxo-7-phenylheptanoate, an intermediate for the synthesis of substituted octahydroindoles.[1]
  • In the preparation of ortho-bromo phenethylamine products, which are further used to synthesize chiral 2-substituted indolines.[2]
  • In the synthesis of β-aryltelluro amines as potent carbonic anhydrase inhibitors.[3]

(S)-(+)-2-Benzyl-1-(p-tolylsulfonyl)aziridine may be used in the preparation of (R)-ethyl (5-benzyl-2-oxo-1-tosyl)pyrrolidine-3-carboxylate by reacting with diethyl potassiomalonate.[4]

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Stereochemical studies. 44. Exploitation of the new synthetic scheme for chiral additives usable in asymmetric syntheses. Novel syntheses of optically active. GAMMA.-amino acids and pyrrolidines from L-a-amino acids.
Tseng CC, et al.
Chemical & Pharmaceutical Bulletin, 25(1), 29-40 (1977)
Asymmetric Synthesis of Octahydroindoles via a Domino Robinson Annulation/5-Endo Intramolecular Aza-Michael Reaction
Parra C, et al.
The Journal of Organic Chemistry, 81(21), 10172-10179 (2016)
First evaluation of organotellurium derivatives as carbonic anhydrase I, II, IV, VII and IX inhibitors
Angeli A, et al.
Bioorganic Chemistry, 76(17), 268-272 (2018)
Ring opening of aziridines with ortho-bromophenyl metal reagents: synthesis of 2-substituted indolines
Michaelis DJ and Dineen TA
Tetrahedron Letters, 50(17), 1920-1923 (2009)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service