Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

463892

Sigma-Aldrich

Gallium(III) sulfate hydrate

≥99.99% trace metals basis

Synonym(s):

Digallium trisulfate hydrate, Gallium sulfate hydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Ga2(SO4)3 · xH2O
CAS Number:
Molecular Weight:
427.63 (anhydrous basis)
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
assay:
≥99.99% trace metals basis
form:
solid

assay

≥99.99% trace metals basis

form

solid

reaction suitability

reagent type: catalyst
core: gallium

impurities

≤100.0 ppm Trace Metal Analysis

mp

105-110 °C (lit.)

density

3.86 g/mL at 25 °C (lit.)

SMILES string

O.[Ga+3].[Ga+3].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O

InChI

1S/2Ga.3H2O4S.H2O/c;;3*1-5(2,3)4;/h;;3*(H2,1,2,3,4);1H2/q2*+3;;;;/p-6

InChI key

CMNGAUGWXGMLDK-UHFFFAOYSA-H

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Stijn Raiguel et al.
Dalton transactions (Cambridge, England : 2003), 49(11), 3532-3544 (2020-02-29)
Two 1,2,3-triazolium ionic liquid diluents were evaluated for the extraction of gallium from spent Bayer process liquor by Kelex® 100. The first of these is a hydrophobic ionic liquid with a low water content, which allows the extraction of gallium

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service