Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

463388

Sigma-Aldrich

2-Hydroxypropyl 2-(methacryloyloxy)ethyl phthalate

Synonym(s):

2-Hydroxypropyl-2-methacryloyloxyethyl phthalate, 2-Methacryloyloxyethyl 2-hydroxypropyl phthalate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
2-[H2C=C(CH3)CO2CH2CH2O2C]C6H4CO2CH2CH(OH)CH3
CAS Number:
Molecular Weight:
336.34
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

contains

900 ppm monomethyl ether hydroquinone as inhibitor

impurities

50% 2-methyl-1-hydroxyethyl 2-(methacryloyloxy)ethyl phthalate

refractive index

n20/D 1.513 (lit.)

density

1.063 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

CC(O)COC(=O)c1ccccc1C(=O)OCCOC(=O)C(C)=C

InChI

1S/C17H20O7/c1-11(2)15(19)22-8-9-23-16(20)13-6-4-5-7-14(13)17(21)24-10-12(3)18/h4-7,12,18H,1,8-10H2,2-3H3

InChI key

GLQNPYLKSCFVPO-UHFFFAOYSA-N

pictograms

CorrosionExclamation mark

signalword

Danger

Hazard Classifications

Eye Dam. 1 - Skin Irrit. 2 - Skin Sens. 1

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

235.4 °F - closed cup

flash_point_c

113 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service