Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

462993

Sigma-Aldrich

Methylsilane

electronic grade, ≥99.9%

Synonym(s):

Monosilylmethane, Silaethane, Silylmethane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3SiH3
CAS Number:
Molecular Weight:
46.14
Beilstein/REAXYS Number:
1730728
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.23

grade

electronic grade

vapor density

0.628 (−58 °C, vs air)

assay

≥99.9%

form

liquid

impurities

<10 ppm Carbon dioxide (CO2)
<10 ppm Nitrogen (N2)
<100 ppm Dimethylsilane (CH3)2SiH2
<100 ppm Methylchlorosilane CH3SiH2Cl
<2 ppm Argon + Oxygen (Ar + O2)
<50 ppm Methane (CH4)
<50 ppm Silane (SiH4)

bp

−57 °C (lit.)

mp

−157 °C (lit.)

SMILES string

C[SiH3]

InChI

1S/CH6Si/c1-2/h1-2H3

InChI key

UIUXUFNYAYAMOE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Features and Benefits

Methylsilane is used in the plasma-assisted CVD of silicon carbide (SiC) and as a precursor to the epitaxial growth of SiGeC alloy layers via rapid thermal CVD.[1] Recent developments include the use of methylsilane as a precursor to silicon-carbon-hydrogen polymer used as photoresists.
Precursor to silicon carbide or alloy thin films by chemical vapor deposition (CVD) and molecular beam epitaxy (MBE).

Recommended products

Stainless steel regulators Z527416 or Z527424 are recommended.

signalword

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Flam. Gas 1 - Press. Gas Liquefied gas

supp_hazards

Storage Class

2A - Gases

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Faceshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yifan Li et al.
Beilstein journal of nanotechnology, 8, 2504-2514 (2017-12-21)
Interface conditions are an important property that can affect the drag of fluid flow. For surfaces with different oleophobicity, the boundary slip at the solid-oil interface is mostly larger than that at the solid-water interface. Roughness is a key factor
Mi, J.
J. Vac. Sci. Technol. B, 14, 1660-1660 (1996)
Hai-Feng Wu et al.
Natural product research, 31(21), 2484-2490 (2017-04-14)
One new cycloartane triterpenoid glycoside, soulieoside Q (1), together with four known compounds (2-5) were isolated from the ethanolic extract of the rhizomes of Souliea vaginata Maxim. The structure of the new compound was determined by extensive spectroscopic analysis including
C A Diaz-Botia et al.
Journal of neural engineering, 14(5), 056006-056006 (2017-06-03)
Current neural probes have a limited device lifetime of a few years. Their common failure mode is the degradation of insulating films and/or the delamination of the conductor-insulator interfaces. We sought to develop a technology that does not suffer from
Zulan Li et al.
Biosensors & bioelectronics, 166, 112444-112444 (2020-08-08)
How to balance the sensitivity and signal-to-noise ratio of immunosensor remains many challenges during various diseases diagnosis. Here we develop a new microfluidic immunosensor based on surface-modified mesoporous nanofibers, and simultaneously realize an ultra-sensitivity and high signal-to-noise ratio for the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service