Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

461423

Sigma-Aldrich

(R)-(+)-N-(1-Phenylethyl)succinamic acid

98%

Synonym(s):

(R)-(+)-N-(α-Methylbenzyl)succinamidic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH(CH3)NHCOCH2CH2CO2H
CAS Number:
Molecular Weight:
221.25
Beilstein/REAXYS Number:
3205512
EC Number:
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:
NACRES:
NA.22

assay

98%

form

solid

optical activity

[α]24/D +111°, c = 2 in ethanol

mp

100-104 °C (lit.)

SMILES string

C[C@@H](NC(=O)CCC(O)=O)c1ccccc1

InChI

1S/C12H15NO3/c1-9(10-5-3-2-4-6-10)13-11(14)7-8-12(15)16/h2-6,9H,7-8H2,1H3,(H,13,14)(H,15,16)/t9-/m1/s1

InChI key

WUEKFTPKHWMMIP-SECBINFHSA-N

Application

Acid for the resolution of amines.[1][2]

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kozma, D. et al.
J. Therm. Anal., 44, 339-339 (1995)
Acs, M. et al.
Chirality, 6, 314-314 (1994)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service