Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

460982

Sigma-Aldrich

N,N-Diethylbutylamine

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)3N(C2H5)2
CAS Number:
Molecular Weight:
129.24
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

97%

refractive index

n20/D 1.414 (lit.)

bp

136.5 °C (lit.)

density

0.748 g/mL at 25 °C (lit.)

functional group

amine

SMILES string

CCCCN(CC)CC

InChI

1S/C8H19N/c1-4-7-8-9(5-2)6-3/h4-8H2,1-3H3

InChI key

ORSUTASIQKBEFU-UHFFFAOYSA-N

General description

N,N-Diethylbutylamine is an N,N-diethylalkylamine.[1] It has been reported to induce carotenogenesis in Phycomyces blakesleeanus wild type NRRL 1555 and carA mutants C2, C3 and C152.[2]

Application

N,N-Diethylbutylamine may be used for the regioselective syntheses of of 6-(1,1-dimethylallyl) ortho-flavones.[1][3] It was used in the synthesis of N,N-diethylbutylammonium bis(trifluoromethylsulfonyl)imide.[4]

pictograms

FlameCorrosion

signalword

Danger

hcodes

Hazard Classifications

Flam. Liq. 3 - Skin Corr. 1B

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

75.2 °F - closed cup

flash_point_c

24 °C - closed cup

ppe

Faceshields, Gloves, Goggles, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Regioselective syntheses of 6-(1, 1-dimethylallyl)-and 8-(3, 3-dimethylallyl) chrysins.
Daskiewicz J-B, et al.
Tetrahedron, 58(18), 3589-3595 (2002)
Chemical bioregulation of carotenogenesis in Phycomyces blakesleeanus.
Hsu W-J, et al.
Phytochemistry, 29(8), 2447-2451 (1990)
Shohei Kakinuma et al.
The journal of physical chemistry. B, 122(22), 6033-6047 (2018-05-11)
In this study, we have investigated the effects of cation structures on the temperature dependence of the intermolecular vibrational dynamics of ionic liquids using femtosecond Raman-induced Kerr effect spectroscopy. The ionic liquids used in this study are bis(trifluoromethylsulfonyl)amide [NTf2]- salts
Rearrangement of 5-O-prenyl flavones: a regioselective access to 6-C-(1,1-dimethylallyl)-and 8-C-(3, 3-dimethylallyl)-flavones.
Daskiewicz J-B, et al.
Tetrahedron Letters, 42(41), 7241-7244 (2001)
Solvent Extraction of Sr2+ and Cs+ Based on Hydrophobic Protic Ionic Liquids
Luo H, et al.
Zeitschrift fur Naturforschung, A, 62(5-6), 281-291 (2007)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service