Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

46084

Sigma-Aldrich

2,2′-(Ethylenedithio)diacetic acid

≥97.0%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H10O4S2
CAS Number:
Molecular Weight:
210.27
Beilstein/REAXYS Number:
1778542
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

≥97.0%

mp

107-110 °C

SMILES string

OC(=O)CSCCSCC(O)=O

InChI

1S/C6H10O4S2/c7-5(8)3-11-1-2-12-4-6(9)10/h1-4H2,(H,7,8)(H,9,10)

InChI key

CCQMORVULNZXIA-UHFFFAOYSA-N

General description

2,2′-(Ethylenedithio)diacetic acid ((Ethylenedithio)diacetic acid) is a linear aliphatic thiopolycarboxylic acid.[1] Polycondensation reaction of 2,2′-(ethylenedithio)diacetic acid has been reported.[2] Protonation constant of ethylenedithiodiacetic acid has been evaluated from potentiometric measurements in NaCl(aq) and (CH3)4NCl(aq) at 25°C. Dependence of protonation constant on ionic strength has been modeled by the SIT and Pitzer approaches.[3]

Application

2,2′-(Ethylenedithio)diacetic acid ((Ethylenedithio)diacetic acid) has been used in the spectrophotometric titration of mercury(II).[1]

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Spectrophotometric titration of mercury (II) with ethylenedithiodiacetic acid.
Napoli A and Paolillo M.
Microchimica Acta, 70(3-4), 297-301 (1978)
Modeling of protonation constants of linear aliphatic dicarboxylates containing-S-groups in aqueous chloride salt solutions, at different ionic strengths, using the SIT and Pitzer equations and empirical relationships.
Bretti C, et al.
Journal of Solution Chemistry, 37(6), 763-784 (2008)
Synthesis of polyanhydrides. XI. 1 Synthesis and properties of new polythioetherpolyanhydrides.
Yoda N.
Makromol. Chem., 56(1), 36-54 (1962)
Mavis D Boamah et al.
The journal of physical chemistry. B, 123(27), 5848-5856 (2019-07-02)
Ion specific outcomes at aqueous interfaces remain among the most enigmatic phenomena in interfacial chemistry. Here, charged fused silica/water interfaces have been probed by homodyne- and heterodyne-detected (HD) second harmonic generation (SHG) spectroscopy at pH 7 and 5.8 and for

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service