Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

460141

Sigma-Aldrich

Poly(ethylene glycol) monolaurate

average Mn ~600

Synonym(s):

Polyethylene glycol 400 monolaurate

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:

mol wt

average Mn ~600

refractive index

n20/D 1.46

bp

>260 °C (lit.)

hydroxyl value

95 mg KOH/g

iodine value

8

solubility

propylene glycol: insoluble
toluene, ethanol, acetone and water: soluble

density

1.029 g/mL at 25 °C

SMILES string

OCCO.CCCCCCCCCCCC(O)=O

InChI

1S/C14H28O3/c1-2-3-4-5-6-7-8-9-10-11-14(16)17-13-12-15/h15H,2-13H2,1H3

InChI key

CTXGTHVAWRBISV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Plasticizer, surfactant.

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

235.4 °F - closed cup

flash_point_c

113 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

V V Dmitriev et al.
Mikrobiologiia, 57(2), 223-230 (1988-03-01)
Yeasts capable of utilizing the non-ionogenic surfactant Laurox-9 were identified as Cryptococcus humicolus (Daszewska) Golubev and Rhodotorula mucilaginosa (Jöerg) Harrison. IR spectrometry used for the quantitative assay of Laurox-9 showed that R. mucilaginosa utilized it as a sole carbon source.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service