Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

459518

Sigma-Aldrich

2,4-Dibromo-1-fluorobenzene

90%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Br2C6H3F
CAS Number:
Molecular Weight:
253.89
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

assay

90%

refractive index

n20/D 1.583 (lit.)

bp

214-216 °C/756 mmHg (lit.)

density

2.047 g/mL at 25 °C (lit.)

SMILES string

Fc1ccc(Br)cc1Br

InChI

1S/C6H3Br2F/c7-4-1-2-6(9)5(8)3-4/h1-3H

InChI key

MKHDDTWHDFVYDQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

197.6 °F - closed cup

flash_point_c

92 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tetrahedron Letters, 37, 6551-6551 (1996)
Pestic. Sci., 28, 25-25 (1990)
T Kayano et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 20(50), 16583-16589 (2014-10-21)
We investigated the luminescence properties and color tuning of [Pt(dpb)Cl] (dpbH=1,3-di(2-pyridyl)benzene) and its analogues. An almost blue emission was obtained for the complex [Pt(Fmdpb)CN] (FmdpbH=4-fluoro-1,3-di(4-methyl-2-pyridyl)benzene), modified by the introduction of F and CH3 groups to the dpb ligand and the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service