Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

459011

Sigma-Aldrich

3,3′-Thiodipropionic acid, polymer-bound

Thiodipropionic acid on DOWEX SBR resin., 20-50 mesh, extent of labeling: ~2.5 mmol/g S loading

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.22

form

solid

reaction suitability

reaction type: solution phase peptide synthesis

extent of labeling

~2.5 mmol/g S loading

particle size

20-50 mesh

SMILES string

OC(=O)CCSCCC(O)=O

InChI

1S/C6H10O4S/c7-5(8)1-3-11-4-2-6(9)10/h1-4H2,(H,7,8)(H,9,10)

InChI key

ODJQKYXPKWQWNK-UHFFFAOYSA-N

Application

For reductive quenching of ozonolysis reactions.[1]
Replacement for volatile dimethyl sulfide. Filtration of reaction solution gives reaction medium free of sulfur-containing by-products.

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Eye Irrit. 2

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Appell, R.B., et al.
Synthetic Communications, 25, 3589-3589 (1995)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service