Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

458988

Sigma-Aldrich

Polyethylene-block-poly(ethylene glycol)

average Mn ~920

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2(CH2CH2)X(OCH2CH2)YOH
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

form

solid

mol wt

average Mn ~920

composition

ethylene oxide, 50 wt. %

CMC

28.8 mg/L (25°C)

surface tension

45 dyn/cm, 25 °C

hydroxyl value

55 mg KOH/g

transition temp

Tm 105 °C (DSC)

solubility

mineral oil, cyclohexane, toluene and MEK: soluble

HLB

10

SMILES string

C=C.OCCO

Looking for similar products? Visit Product Comparison Guide

Application

Nonionic surfactant.
Nonionic surfactant.
Emulsifier for coatings, ceramics, metalworking fluids and inks. Lubricant. Mold release agent. Thickening agent.

Features and Benefits

Polyether segment is susceptible to oxidative degradation.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service