Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

458813

Sigma-Aldrich

Thallium(I) iodide

anhydrous, beads, −10 mesh, 99.999% trace metals basis

Synonym(s):

Thallium iodide, Thallium monoiodide, Thallous iodide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
TlI
CAS Number:
Molecular Weight:
331.29
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:

grade

anhydrous

assay

99.999% trace metals basis

form

beads

particle size

−10 mesh

mp

440 °C (lit.)

density

7.29 g/mL at 25 °C (lit.)

SMILES string

I[Tl]

InChI

1S/HI.Tl/h1H;/q;+1/p-1

InChI key

CMJCEVKJYRZMIA-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

signalword

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Aquatic Chronic 2 - STOT RE 2

Storage Class

6.1B - Non-combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service