458716
Poly[di(ethylene glycol)/cyclohexanedimethanol-alt-isophthalic acid, sulfonated]
average Mn ~8,000 (GPC vs. polyethylene stds.)
About This Item
Recommended Products
mol wt
average Mn ~8,000 (GPC vs. polyethylene stds.)
composition
sulfonate groups, 5-8 mol %
acid number
<2 mg KOH/g
hydroxyl value
<10 mg KOH/g
transition temp
Tg (DSC) 52 °C (onset)
solubility
cyclohexane, diisopropyl ketone, and 2-ethylhexanol: insoluble
water, methylene chloride and 1,6-hexanediol: soluble
density
1.2 g/mL at 25 °C (lit.)
Application
Physical form
Storage Class
13 - Non Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service