Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

458430

Sigma-Aldrich

Hafnium(IV) iodide

anhydrous, 99%

Synonym(s):

Hafnium tetraiodide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HfI4
CAS Number:
Molecular Weight:
686.11
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
assay:
99%
grade:
anhydrous
form:
powder

grade

anhydrous

Quality Level

assay

99%

form

powder

density

5.6 g/mL at 25 °C (lit.)

SMILES string

I[Hf](I)(I)I

InChI

1S/Hf.4HI/h;4*1H/q+4;;;;/p-4

InChI key

YCJQNNVSZNFWAH-UHFFFAOYSA-J

pictograms

Health hazard

signalword

Danger

hcodes

Hazard Classifications

Repr. 1B

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type P2 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service