Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

458422

Sigma-Aldrich

Poly(1,6-hexanediol/neopentyl glycol-alt-adipic acid)

average Mn ~900

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12162002

form

solid

mol wt

average Mn ~900

refractive index

n20/D 1.468 (lit.)

viscosity

330 cP(60 °C)(lit.)

bp

>350 °C (lit.)

hydroxyl functionality

2.0

hydroxyl value

120 mg KOH/g

transition temp

Tm 33 °C (DSC, peak)

density

1.05 g/mL at 25 °C (lit.)

InChI

1S/C6H10O4.C6H14O2.C5H12O2/c7-5(8)3-1-2-4-6(9)10;7-5-3-1-2-4-6-8;1-5(2,3-6)4-7/h1-4H2,(H,7,8)(H,9,10);7-8H,1-6H2;6-7H,3-4H2,1-2H3

InChI key

HNIBILYKBWAIPI-UHFFFAOYSA-N

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service