Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

458104

Sigma-Aldrich

1-Chloro-3,3,4,4,5,5-hexafluoro-2-methoxycyclopentene

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H3ClF6O
CAS Number:
Molecular Weight:
240.53
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

assay

97%

refractive index

n20/D 1.376 (lit.)

bp

84 °C (lit.)

density

1.56 g/mL at 25 °C (lit.)

SMILES string

COC1=C(Cl)C(F)(F)C(F)(F)C1(F)F

InChI

1S/C6H3ClF6O/c1-14-3-2(7)4(8,9)6(12,13)5(3,10)11/h1H3

InChI key

QLPINEFCFXPUSF-UHFFFAOYSA-N

Application

Used in the preparation of perhalocyclopentanones and more recently to prepare tetracarbocyanine dyes with fluorinated rings in the side chain.

pictograms

FlameExclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

120.2 °F - closed cup

flash_point_c

49 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The Journal of Organic Chemistry, 33, 2692-2692 (1968)
Zh. Org. Khim., 28, 1508-1508 (1992)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service