Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

457205

Sigma-Aldrich

Poly(styrene-co-α-methylstyrene)

melt viscosity 100 poise (161 °C)

Synonym(s):

α-Methylstyrene-styrene, α-Methylstyrene-styrene polymer, Styrene-α-methylstyrene copolymer, Styrene-α-methylstyrene resin

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[CH2CH(C6H5)]x[CH2C(CH3)(C6H5)]y
CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

form

solid

melt viscosity

100 poise(161 °C)

acid number

<1 mg KOH/g

transition temp

softening point 119 °C (ring and ball)
Tg (DSC) 67 °C (onset)

solubility

hydrocarbons, esters and ketones: soluble
water, alcohols and glycols: insoluble

density

1.07 g/mL at 25 °C

SMILES string

C=Cc1ccccc1.CC(=C)c2ccccc2

InChI

1S/C9H10.C8H8/c1-8(2)9-6-4-3-5-7-9;1-2-8-6-4-3-5-7-8/h3-7H,1H2,2H3;2-7H,1H2

InChI key

ZAKVZVDDGSFVRG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Additive to sealants, caulks, adhesives, and coatings.

Features and Benefits

Compatible with acrylic, styrene, butadiene and rosin polymers.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service