Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

456829

Sigma-Aldrich

Cerium(III) iodide

anhydrous, powder, ≥99.95%

Synonym(s):

Cerous triiodide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CeI3
CAS Number:
Molecular Weight:
520.83
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:

grade

anhydrous

assay

≥99.95%

form

powder

reaction suitability

reagent type: catalyst
core: cerium

mp

750 °C (lit.)

SMILES string

I[Ce](I)I

InChI

1S/Ce.3HI/h;3*1H/q+3;;;/p-3

InChI key

ZEDZJUDTPVFRNB-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Faceshields, Gloves, type P2 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service