Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

456721

Sigma-Aldrich

tert-Butyllithium solution

1.5 M in pentane

Synonym(s):

Lithium-2-methyl-2-propanide, t-BuLi

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3CLi
CAS Number:
Molecular Weight:
64.06
Beilstein/REAXYS Number:
3587204
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
Pricing and availability is not currently available.

concentration

1.5 M in pentane

storage temp.

2-8°C

SMILES string

[Li]C(C)(C)C

InChI

1S/C4H9.Li/c1-4(2)3;/h1-3H3;

InChI key

BKDLGMUIXWPYGD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Packaging

Packaged under nitrogen in Sure/Seal bottles

Legal Information

Sure/Seal is a trademark of Sigma-Aldrich Co. LLC

signalword

Danger

Hazard Classifications

Aquatic Chronic 2 - Asp. Tox. 1 - Eye Dam. 1 - Flam. Liq. 2 - Pyr. Liq. 1 - Skin Corr. 1B - STOT SE 3 - Water-react 1

target_organs

Central nervous system

Storage Class

4.2 - Pyrophoric and self-heating hazardous materials

wgk_germany

WGK 2

flash_point_f

-40.0 °F

flash_point_c

-40 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xinyi Chia et al.
ACS nano, 9(5), 5164-5179 (2015-04-22)
Layered transition metal dichalcogenides (TMDs) have been the center of attention in the scientific community due to their properties that can be tapped on for applications in electrochemistry and hydrogen evolution reaction (HER) catalysis. We report on the effect of
Adeline Huiling Loo et al.
Chemphyschem : a European journal of chemical physics and physical chemistry, 16(11), 2304-2306 (2015-05-28)
Owing to the attractive properties that transition metal dichalcogenides (TMDs) display, they have found recent application in the fabrication of biosensing devices. These devices involve the immobilization of a recognition element such as DNA onto the surface of TMDs. Therefore
Shigeo Hayashi et al.
Journal of enzyme inhibition and medicinal chemistry, 29(6), 846-867 (2014-02-13)
Because of the pivotal role of cyclooxygenase (COX) in the inflammatory processes, non-steroidal anti-inflammatory drugs (NSAIDs) that suppress COX activities have been used clinically for the treatment of inflammatory diseases/syndromes; however, traditional NSAIDs exhibit serious side-effects such as gastrointestinal damage

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service