Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

456497

Sigma-Aldrich

Benzyl (R)-(−)-mandelate

99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH(OH)CO2CH2C6H5
CAS Number:
Molecular Weight:
242.27
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:
NACRES:
NA.22

assay

99%

form

solid

optical activity

[α]25/D −55°, c = 1 in chloroform

mp

104-107 °C (lit.)

SMILES string

O[C@@H](C(=O)OCc1ccccc1)c2ccccc2

InChI

1S/C15H14O3/c16-14(13-9-5-2-6-10-13)15(17)18-11-12-7-3-1-4-8-12/h1-10,14,16H,11H2/t14-/m1/s1

InChI key

JFKWZVQEMSKSBU-CQSZACIVSA-N

Application

Benzyl (R)-(-)-mandelate can be employed as reactant to synthesize:
  • (S)-α-Hydroxy-N-(2-phenylethyl)benzenepropanamide (α-hydroxyamides) by enzyme catalyzed amidation reaction.[1]
  • Optically pure (R)- monomethyl esters of 3-[(tert-butyldimethylsilyl)oxy]pentanedioic acid, which is employed in the preparation of HMG-CoA reductase inhibitors.[2]
  • O-Ibuprofenylmandelic acid via acylation reaction with optically active ibuprofen and subsequent hydrogenation.[3]
  • Benzyl (R)-2-fluoro-2-phenylacetate via deoxyfluorination of alcohols using AlkylFluor.[4]
It can also be used to prepare HMG-CoA reductase inhibitors.[5]

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Practical synthesis of chiral synthons for the preparation of HMG-CoA reductase inhibitors
Konoike T and Araki Y
The Journal of Organic Chemistry, 59(25), 7849-7854 (1994)
Konoike, T. Araki, Y.
The Journal of Organic Chemistry, 59, 7849-7849 (1994)
Preparation of optically active (acyloxy) alkyl esters from optically active O-acyl-α-hydroxy acids
Guzzo Peter R, et al.
Tetrahedron Letters, 43(32), 5685-5689 (2002)
Stereoselective Pseudomonas cepacia lipase mediated synthesis of α-hydroxyamides
Adamczyk M, et al.
Tetrahedron Asymmetry, 8(15), 2509-2512 (1997)
AlkylFluor: Deoxyfluorination of Alcohols.
Goldberg NW, et al.
Organic Letters, 18(23), 6102-6104 (2016)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service