Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

454842

Sigma-Aldrich

N-Acetylphthalimide

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H7NO3
CAS Number:
Molecular Weight:
189.17
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

assay

97%

mp

139 °C (lit.)

SMILES string

CC(=O)N1C(=O)c2ccccc2C1=O

InChI

1S/C10H7NO3/c1-6(12)11-9(13)7-4-2-3-5-8(7)10(11)14/h2-5H,1H3

Inchi Key

INZUQGFQRYAKQQ-UHFFFAOYSA-N

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rhodium-Catalyzed Synthesis and Reactions of N-Acylphthalimides.
Li G, et al.
Asian Journal of Organic Chemistry, 2(11), 983-988 (2013)
Structure and Synthesis of the Major Components in the Hairpencil Secretion of a Male Butterfly, Lycorea ceres ceres (Cramer) 1.
Meinwald J and Meinwald YC.
Journal of the American Chemical Society, 88(6), 1305-1310 (1966)
The hydrolysis and cyclization of some phthalamic acid derivatives.
J Brown et al.
Journal of the American Chemical Society, 88(19), 4468-4474 (1966-10-05)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service