Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

454117

Sigma-Aldrich

Phenyl propargyl sulfide

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5SCH2C≡CH
CAS Number:
Molecular Weight:
148.22
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

97%

refractive index

n20/D 1.593 (lit.)

bp

74 °C/0.4 mmHg (lit.)

density

1.077 g/mL at 25 °C (lit.)

SMILES string

C#CCSc1ccccc1

InChI

1S/C9H8S/c1-2-8-10-9-6-4-3-5-7-9/h1,3-7H,8H2

InChI key

HXKVNRKJSSHQQY-UHFFFAOYSA-N

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

208.4 °F - closed cup

flash_point_c

98 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Highly efficient [2, 3]-sigmatropic rearrangement of sulfur ylide derived from Rh (II) carbene and sulfides in water.
Liao M and Wang J.
Green Chemistry, 9(2), 184-188 (2007)
Transition-metal-catalyzed rearrangement of allenyl sulfides: a route to furan derivatives.
Lingling Peng et al.
Angewandte Chemie (International ed. in English), 46(11), 1905-1908 (2007-02-03)
Synthesis and characterization of sulfur-containing polyacetylene derivative: Synthesis of poly (phenyl propargyl sulfide) and its electro-optical properties.
Gal Y-S, et al.
Current Applied Physics, 7(5), 480-484 (2007)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service