Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

45366

Sigma-Aldrich

(S)-(−)-1,2-Epoxyoctane

≥98.0% (sum of enantiomers, GC)

Synonym(s):

(S)-(−)-1-Octene oxide, (S)-(−)-2-Hexyloxirane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H16O
CAS Number:
Molecular Weight:
128.21
Beilstein/REAXYS Number:
79914
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:

assay

≥98.0% (sum of enantiomers, GC)

optical activity

[α]20/D −14.5±1°, c = 2.5% in ethanol

optical purity

enantiomeric ratio: ≥98.5:1.5 (HPLC)

refractive index

n20/D 1.421

density

0.835 g/mL at 20 °C (lit.)

storage temp.

2-8°C

SMILES string

CCCCCC[C@H]1CO1

InChI

1S/C8H16O/c1-2-3-4-5-6-8-7-9-8/h8H,2-7H2,1H3/t8-/m0/s1

InChI key

NJWSNNWLBMSXQR-QMMMGPOBSA-N

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

pictograms

FlameExclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

98.6 °F - closed cup

flash_point_c

37 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kelli L Hvorecny et al.
Structure (London, England : 1993), 25(5), 697-707 (2017-04-11)
Pseudomonas aeruginosa secretes an epoxide hydrolase with catalytic activity that triggers degradation of the cystic fibrosis transmembrane conductance regulator (CFTR) and perturbs other host defense networks. Targets of this CFTR inhibitory factor (Cif) are largely unknown, but include an epoxy-fatty

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service