Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

453420

Sigma-Aldrich

(1S,2R,4S)-(+)-4-(1-Methylethyl)-2-(1-methyl-1-phenylethyl)cyclohexanol

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHC6H9[C(CH3)2C6H5]OH
UNSPSC Code:
12352002
PubChem Substance ID:
NACRES:
NA.22

assay

98%

optical activity

[α]20/D +35°, c = 1 in ethanol

optical purity

ee: 99% (HPLC)

mp

68-71 °C (lit.)

SMILES string

CC(C)[C@H]1CC[C@H](O)[C@H](C1)C(C)(C)c2ccccc2

InChI

1S/C18H28O/c1-13(2)14-10-11-17(19)16(12-14)18(3,4)15-8-6-5-7-9-15/h5-9,13-14,16-17,19H,10-12H2,1-4H3/t14-,16-,17-/m0/s1

InChI key

ZJHNUUVRRFRSFM-XIRDDKMYSA-N

Application

Cyclohexyl-based chiral auxiliary that provides equal or greater levels of asymmetric induction when compared to menthol or 8-phenylmenthol in a variety of reactions.[1][2][3][4]

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Comins, D.L. Guerra-Weltzien, L.
Tetrahedron Letters, 37, 3807-3807 (1996)
Comins, D.L. et al.
Journal of the American Chemical Society, 116, 4719-4719 (1994)
Comins, D.L. et al.
Synlett, 972-972 (1994)
Comins, D.L. Radi Benjelloun, N.
Tetrahedron Letters, 35, 829-829 (1994)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service