Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

452769

Sigma-Aldrich

1-Iodo-3,5-dinitrobenzene

new

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
IC6H3(NO2)2
CAS Number:
Molecular Weight:
294.00
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

assay

98%

form

crystals

mp

108-111 °C (lit.)

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

[O-][N+](=O)c1cc(I)cc(c1)[N+]([O-])=O

InChI

1S/C6H3IN2O4/c7-4-1-5(8(10)11)3-6(2-4)9(12)13/h1-3H

InChI key

AISNAASNOWRWIR-UHFFFAOYSA-N

General description

1-Iodo-3,5-dinitrobenzene, also known as 3,5-dinitroiodobenzene, is generally used in dyes, stains, indicators, general laboratory use, and other industrial applications. It can also be used to synthesize 1-iodo-3,5-diaminobenzene by the reduction of nitro groups.[1] Diaminobenzenes are key intermediates in the preparation of stains and dyes.[2]

pictograms

Exclamation mark

signalword

Warning

hcodes

pcodes

Hazard Classifications

Skin Sens. 1

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Faceshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of carbon dendron nano-chains with ?-extended conjugation based on linear 1, 4-phenylethynyl and 1, 5-naphthylethynyl subunits
Rodriguez JG, et al.
Tetrahedron Letters, 44(34), 6375-6378 (2003)
Amines, Aromatic, Diarylamines
Layer RW
Kirk-Othmer Encyclopedia of Chemical Technology, 44(34), 6375-6378 (2000)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service