452149
Polypropylene
Syndiotactic, average Mw ~174,000, average Mn ~75,000
About This Item
Recommended Products
gloss
51.6 % (45°C)
melt index
2.2 g/10 min (230°C/2.16kg)
mol wt
average Mn ~75,000
average Mw ~174,000
impact strength
14.9 ft-lb/in. (Izod)
transition temp
Tm 125 °C (DSC, peak)
density
0.9 g/mL at 25 °C (lit.)
SMILES string
CC=C
InChI
1S/C22H42O3/c1-2-3-4-5-11-14-17-20-21(25-20)18-15-12-9-7-6-8-10-13-16-19-22(23)24/h20-21H,2-19H2,1H3,(H,23,24)/t20-,21+/m1/s1
InChI key
NSYDMBURIUSUDH-RTWAWAEBSA-N
Looking for similar products? Visit Product Comparison Guide
Physical form
Storage Class
13 - Non Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service