Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

451673

Sigma-Aldrich

Antimony(V) chloride

anhydrous, ≥99.999% trace metals basis

Synonym(s):

Antimony pentachloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
SbCl5
CAS Number:
Molecular Weight:
299.03
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:

grade

anhydrous

vapor density

>10.2 (vs air)

vapor pressure

0.13 psi ( 55 °C)

assay

≥99.999% trace metals basis

form

liquid

reaction suitability

reagent type: catalyst
core: antimony

bp

92 °C/30 mmHg (lit.)

mp

2.8 °C (lit.)

density

2.36 g/mL at 25 °C (lit.)

SMILES string

Cl[Sb](Cl)(Cl)(Cl)Cl

InChI

1S/5ClH.Sb/h5*1H;/q;;;;;+5/p-5

InChI key

VMPVEPPRYRXYNP-UHFFFAOYSA-I

Looking for similar products? Visit Product Comparison Guide

signalword

Danger

Hazard Classifications

Aquatic Chronic 2 - Eye Dam. 1 - Ox. Liq. 2 - Skin Corr. 1B - STOT SE 3

target_organs

Respiratory system

Storage Class

5.1B - Oxidizing hazardous materials

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Faceshields, Gloves, Goggles, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

In-transit chemical spill--West Virginia.
MMWR. Morbidity and mortality weekly report, 35(6), 81-82 (1986-02-14)
M P Oliveira-Neto et al.
Ophthalmology, 107(1), 169-172 (2000-01-27)
To describe American cutaneous leishmaniasis of the eyelids and highlight the main clinical and diagnostic features of lesions, which are rare in this location. Retrospective, noncomparative case series Leishmanin skin test, touch preparations, histopathologic analysis, and culture in appropriate media
J Carrió et al.
Experimental parasitology, 95(3), 209-214 (2000-08-31)
The determination of the intrinsic sensitivity of Leishmania strains to pentavalent antimonials in clinical trials, before treatment is begun, is essential in order to avoid failures and to allow alternative drugs to be chosen. A comparative study of SbV activity
Flávio José Dutra de Moura et al.
Toxicology, 243(1-2), 66-74 (2007-11-21)
Tumor necrosis factor-alpha (TNF-alpha) is a mediator of inflammation and has an important role in human and experimental renal diseases. Pentoxifylline (PTX) has been shown to inhibit cytokine synthesis, including TNF-alpha. The aim of the present study was to examine
Et2SBrSbCl5Br: an effective reagent for direct bromonium-induced polyene cyclizations.
Scott A Snyder et al.
Angewandte Chemie (International ed. in English), 48(42), 7899-7903 (2009-09-17)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service