Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

451606

Sigma-Aldrich

Titanium(IV) bromide

anhydrous, powder, 99.99% trace metals basis

Synonym(s):

Titanium tetrabromide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
TiBr4
CAS Number:
Molecular Weight:
367.48
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:

grade

anhydrous

assay

99.99% trace metals basis

form

powder

bp

230 °C (lit.)

mp

38-40 °C (lit.)

density

3.25 g/mL at 25 °C (lit.)

SMILES string

Br[Ti](Br)(Br)Br

InChI

1S/4BrH.Ti/h4*1H;/q;;;;+4/p-4

InChI key

UBZYKBZMAMTNKW-UHFFFAOYSA-J

Looking for similar products? Visit Product Comparison Guide

Features and Benefits

Used for the formation of titanium silicide thin films on Si substrates.

pictograms

Corrosion

signalword

Danger

hcodes

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

supp_hazards

Storage Class

8A - Combustible corrosive hazardous materials

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Faceshields, Gloves, type P3 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service