Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

451541

Sigma-Aldrich

2,2′,3,3′,5,5′-Hexamethyl-[1,1′-biphenyl]-4,4′-diol

99%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H22O2
CAS Number:
Molecular Weight:
270.37
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:
Pricing and availability is not currently available.

assay

99%

SMILES string

Cc1cc(c(C)c(C)c1O)-c2cc(C)c(O)c(C)c2C

InChI

1S/C18H22O2/c1-9-7-15(11(3)13(5)17(9)19)16-8-10(2)18(20)14(6)12(16)4/h7-8,19-20H,1-6H3

InChI key

IOJCFCLZQBXCIQ-UHFFFAOYSA-N


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service