Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

451223

Sigma-Aldrich

Rubidium chloride

anhydrous, beads, −10 mesh, 99.5% trace metals basis

Synonym(s):

Rubidium monochloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
RbCl
CAS Number:
Molecular Weight:
120.92
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:

grade

anhydrous

assay

99.5% trace metals basis

form

beads

particle size

−10 mesh

mp

715 °C (lit.)

density

2.8 g/mL at 25 °C (lit.)

SMILES string

[Cl-].[Rb+]

InChI

1S/ClH.Rb/h1H;/q;+1/p-1

Inchi Key

FGDZQCVHDSGLHJ-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yucheng Jiang et al.
Carbohydrate research, 341(2), 262-269 (2005-12-07)
The Gibbs energy interaction parameters of RbCl with some monosaccharides (D-glucose, D-galactose, D-xylose, and D-arabinose) in water, g(ES), were obtained from electromotive force (emf) measurements of the electrochemical cell without liquid junction and containing two ion-selective electrodes (ISE): K-ISEmid R:RbCl(m(E))mid
Christophe Danelon et al.
Biophysical chemistry, 104(3), 591-603 (2003-08-14)
Ion current through single outer membrane protein F (OmpF) trimers was recorded and compared to molecular dynamics simulation. Unidirectional insertion was revealed from the asymmetry in channel conductance. Single trimer conductance showed particularly high values at low symmetrical salt solution.
Jerzy Karczewski et al.
Biochemical pharmacology, 77(10), 1602-1611 (2009-05-12)
Drug-induced long QT syndrome has been principally ascribed to block of the cardiac hERG K(+) channel. Methanesulfonanilides, such as MK-499, E-4031 and dofetilide, are potent hERG antagonists that likely bind along the S6 helix within the inner vestibule of the
F Stevens et al.
Physical chemistry chemical physics : PCCP, 7(2), 240-249 (2005-01-21)
An extensive level of theory study is performed on diatomic chalcogen defects in alkali halide lattices by density functional theory methods. A variety of exchange correlation functionals and basis sets are used for the calculation of electron paramagnetic resonance (EPR)
Yui-May Hsiao et al.
Nuclear medicine and biology, 36(1), 39-45 (2009-02-03)
In locations that lack nearby cyclotron facilities for radionuclide production, generator-based (68)Ga radiopharmaceuticals might have clinical utility for positron emission tomography (PET) studies of myocardial perfusion and other physiological processes. The lipophilic and monocationic (67)Ga-labeled gallium chelates of five novel

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service