Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

450332

Sigma-Aldrich

1,2-Phenylenebisphosphine

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H4(PH2)2
CAS Number:
Molecular Weight:
142.08
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:

Quality Level

assay

98%

reaction suitability

reagent type: ligand

bp

53-55 °C/0.25 mmHg (lit.)

density

1.101 g/mL at 25 °C (lit.)

functional group

phosphine

SMILES string

Pc1ccccc1P

InChI

1S/C6H8P2/c7-5-3-1-2-4-6(5)8/h1-4H,7-8H2

InChI key

DKFDVEXWZZOMGS-UHFFFAOYSA-N

pictograms

Flame

signalword

Danger

hcodes

Hazard Classifications

Pyr. Liq. 1

Storage Class

4.2 - Pyrophoric and self-heating hazardous materials

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service